ENA341291067 Enamine

4-isocyanatocyclohex-1-ene

  • MDL number MFCD00050490
  • Cas Number : 4103-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84913
Availability details for
Quantity
  • Availability
  • Add to Cart