ENA341291071 Enamine

1-chloro-2-isothiocyanato-4-methoxybenzene

  • MDL number MFCD04221638
  • Cas Number : 82401-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84969
Availability details for
Quantity
  • Availability
  • Add to Cart