ENA341291088 Enamine

N'-hydroxyfuran-2-carboximidamide

  • MDL number MFCD01726088
  • Cas Number : 50892-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-27100
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart