ENA341291093 Enamine

2-chloro-4-(3-chloropropanamido)benzamide

  • MDL number MFCD12125312
  • Cas Number : 1155902-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-85333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85333
Availability details for
Quantity
  • Availability
  • Add to Cart