ENA341291094 Enamine

4-fluoro-3-sulfanylbenzoic acid

  • MDL number MFCD20441531
  • Cas Number : 1219001-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85334
Availability details for
Quantity
  • Availability
  • Add to Cart