ENA341291102 Enamine

2-[6-(ethanesulfonyl)-2-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl]acetic acid

  • MDL number MFCD09739610
  • Cas Number : 954263-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85477
Availability details for
Quantity
  • Availability
  • Add to Cart