ENA341291106 Enamine

1-(2,4-difluorophenyl)-5-(propan-2-yl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD12557702
  • Cas Number : 1096911-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85545
Availability details for
Quantity
  • Availability
  • Add to Cart