ENA341291110 Enamine

5-amino-1-ethyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD04973471
  • Cas Number : 4788-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85607
Availability details for
Quantity
  • Availability
  • Add to Cart