ENA341291116 Enamine

6-bromo-3-hydroxy-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD11149373
  • Cas Number : 1021021-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85663

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85663
Availability details for
Quantity
  • Availability
  • Add to Cart