ENA341291119 Enamine

ethyl 4-(hydroxymethyl)-1,3-thiazole-2-carboxylate

  • MDL number MFCD12138024
  • Cas Number : 1155532-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85696
Availability details for
Quantity
  • Availability
  • Add to Cart