ENA341291122 Enamine

2-amino-1-(2-bromophenyl)ethan-1-one hydrochloride

  • MDL number MFCD09787500
  • Cas Number : 876063-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85789

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85789
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart