ENA341291124 Enamine

tert-butyl N-{[5-(pyridin-3-yl)-4H-1,2,4-triazol-3-yl]methyl}carbamate

  • MDL number MFCD20441535
  • Cas Number : 1352496-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85805
Availability details for
Quantity
  • Availability
  • Add to Cart