ENA341291130 Enamine

4,5-dichloropyridin-2-amine

  • MDL number MFCD07437842
  • Cas Number : 188577-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85867
Availability details for
Quantity
  • Availability
  • Add to Cart