ENA341291134 Enamine

methyl 2-(4-aminophenyl)-2-methylpropanoate

  • MDL number MFCD20441536
  • Cas Number : 54815-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85928
Availability details for
Quantity
  • Availability
  • Add to Cart