ENA341291153 Enamine

2,5-dimethyl-3-phenylpyrrolidine hydrochloride

  • MDL number MFCD20441540
  • Cas Number : 91562-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86128

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86128
Availability details for
Quantity
  • Availability
  • Add to Cart