ENA341291160 Enamine

(2-chlorophenyl)(4-fluoro-3-methylphenyl)methanamine hydrochloride

  • MDL number MFCD20441543
  • Cas Number : 1354954-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 289...291
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86142
Availability details for
Quantity
  • Availability
  • Add to Cart