ENA341291162 Enamine

3-(2-tert-butyl-5-methylphenoxy)-2-(cyclopropylamino)-2-methylpropanenitrile

  • MDL number MFCD20441545
  • Cas Number : 1306548-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86146

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86146
Availability details for
Quantity
  • Availability
  • Add to Cart