ENA341291178 Enamine

5-fluoro-1-benzofuran-2-carbaldehyde

  • MDL number MFCD20441549
  • Cas Number : 140382-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86182
Availability details for
Quantity
  • Availability
  • Add to Cart