ENA341291182 Enamine

4-(prop-2-yn-1-yloxy)benzoic acid

  • MDL number MFCD00460744
  • Cas Number : 21926-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-213
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86189

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P272,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P333+P313,P337+P313,P362,P363,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-86189
pictograms GHS07
hstatements H302,H315,H317,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart