ENA341291191 Enamine

4-[4-(trifluoromethyl)phenyl]pyridin-3-amine

  • MDL number MFCD20441550
  • Cas Number : 1354962-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86290
Availability details for
Quantity
  • Availability
  • Add to Cart