ENA341291194 Enamine

N-[2-(aminomethyl)phenyl]acetamide hydrochloride

  • MDL number MFCD20441551
  • Cas Number : 238428-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86295
Availability details for
Quantity
  • Availability
  • Add to Cart