ENA341291195 Enamine

5-ethyl-1-(2-fluorophenyl)-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106523
  • Cas Number : 1154370-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86296
Availability details for
Quantity
  • Availability
  • Add to Cart