ENA341291198 Enamine

2,2,2-trifluoroethyl N-(2,4-dichlorophenyl)carbamate

  • MDL number MFCD11099831
  • Cas Number : 1087798-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86300
Availability details for
Quantity
  • Availability
  • Add to Cart