ENA341291200 Enamine

3-(4-aminophenoxy)pyrazine-2-carbonitrile

  • MDL number MFCD16842961
  • Cas Number : 1250251-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86302
Availability details for
Quantity
  • Availability
  • Add to Cart