ENA341291202 Enamine

3-(2-methylpropoxy)cyclohex-2-en-1-one

  • MDL number MFCD00143170
  • Cas Number : 23074-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86304
Availability details for
Quantity
  • Availability
  • Add to Cart