ENA341291205 Enamine

N-methyl-N-(2,2,2-trifluoroethyl)sulfamoyl chloride

  • MDL number MFCD12799606
  • Cas Number : 104468-13-5
  • Molecular Weight : 211.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl(2,2,2-trifluoroethyl)sulfamoyl chloride
Inchi Code 1S/C3H5ClF3NO2S/c1-8(11(4,9)10)2-3(5,6)7/h2H2,1H3
Inchi Key GSWQBHLLDZEMBB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-86307

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-86307
pictograms GHS05,GHS07
hstatements H227,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart