ENA341291213 Enamine

5-(1-chloroethyl)-3-[(4-chlorophenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD10691822
  • Cas Number : 1152532-62-1
  • Molecular Weight : 257.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86319
Availability details for
Quantity
  • Availability
  • Add to Cart