ENA341291218 Enamine

1,2-dimethylpiperidin-4-amine

  • MDL number MFCD12024734
  • Cas Number : 89850-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86329
Availability details for
Quantity
  • Availability
  • Add to Cart