ENA341291225 Enamine

1-(3,5-dimethyl-1,2-oxazole-4-sulfonamido)cyclopentane-1-carboxylic acid

  • MDL number MFCD12542177
  • Cas Number : 1094472-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86341
Availability details for
Quantity
  • Availability
  • Add to Cart