ENA341291227 Enamine

(2-aminoethyl)(3-methoxypropyl)methylamine

  • MDL number MFCD12807109
  • Cas Number : 911300-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86344

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86344
Availability details for
Quantity
  • Availability
  • Add to Cart