ENA341291232 Enamine

2-(aminomethyl)-3-methylbutanoic acid hydrochloride

  • MDL number MFCD20441553
  • Cas Number : 243857-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86355
Availability details for
Quantity
  • Availability
  • Add to Cart