ENA341291233 Enamine

5-formyl-2-methoxybenzonitrile

  • MDL number MFCD18394185
  • Cas Number : 21962-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86390
Availability details for
Quantity
  • Availability
  • Add to Cart