ENA341291243 Enamine

5-chloro-2-(methanesulfonylmethyl)aniline

  • MDL number MFCD16679736
  • Cas Number : 1250988-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86415
Availability details for
Quantity
  • Availability
  • Add to Cart