ENA341291249 Enamine

1-(4-chlorophenyl)-2-methylbutan-1-amine

  • MDL number MFCD11127120
  • Cas Number : 81880-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86423
Availability details for
Quantity
  • Availability
  • Add to Cart