ENA341291251 Enamine

ethyl 2-amino-6-methyl-5,6,7,8-tetrahydro-1,6-naphthyridine-3-carboxylate

  • MDL number MFCD18257056
  • Cas Number : 260247-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86435
Availability details for
Quantity
  • Availability
  • Add to Cart