ENA341291252 Enamine

3-[2-(trifluoromethyl)phenyl]but-2-enoic acid

  • MDL number MFCD11172861
  • Cas Number : 1356809-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86438

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86438
Availability details for
Quantity
  • Availability
  • Add to Cart