ENA341291262 Enamine

1-methyl-4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-7-carboxylic acid

  • MDL number MFCD16085761
  • Cas Number : 1247704-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86472
Availability details for
Quantity
  • Availability
  • Add to Cart