ENA341291264 Enamine

1-[3-(trifluoromethyl)phenyl]cyclopropane-1-carbonitrile

  • MDL number MFCD07374416
  • Cas Number : 124305-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86475
Availability details for
Quantity
  • Availability
  • Add to Cart