ENA341291266 Enamine

1-(pyridine-3-sulfonyl)piperidine-4-carbohydrazide

  • MDL number MFCD20441561
  • Cas Number : 1354950-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86478
Availability details for
Quantity
  • Availability
  • Add to Cart