ENA341291269 Enamine

methyl(1-phenylbutan-2-yl)amine

  • MDL number MFCD12154195
  • Cas Number : 84952-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-86483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86483
Availability details for
Quantity
  • Availability
  • Add to Cart