ENA341291293 Enamine

2-isocyanato-3-methylfuran

  • MDL number MFCD09879906
  • Cas Number : 921938-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86518
Availability details for
Quantity
  • Availability
  • Add to Cart