ENA341291336 Enamine

1-{4-[(2,2,2-trifluoroethane)sulfonyl]phenyl}ethan-1-one

  • MDL number MFCD20441576
  • Cas Number : 1354960-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86563
Availability details for
Quantity
  • Availability
  • Add to Cart