ENA341291337 Enamine

1-[4-(2,2-difluoroethanesulfonyl)phenyl]ethan-1-one

  • MDL number MFCD20441577
  • Cas Number : 1354952-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86564
Availability details for
Quantity
  • Availability
  • Add to Cart