ENA341291354 Enamine

[(3-bromo-2-methoxyphenyl)methyl](methyl)amine

  • MDL number MFCD14656520
  • Cas Number : 1247375-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86582
Availability details for
Quantity
  • Availability
  • Add to Cart