ENA341291356 Enamine

4-(methoxymethyl)benzaldehyde

  • MDL number MFCD11553601
  • Cas Number : 93943-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86584
Availability details for
Quantity
  • Availability
  • Add to Cart