ENA341291392 Enamine

N-(4-methylbenzyl)piperidine-4-carboxamide

  • MDL number MFCD03991892
  • Cas Number : 884497-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86621
Availability details for
Quantity
  • Availability
  • Add to Cart