ENA341291410 Enamine

1-[(2-methylpiperidin-1-yl)sulfonyl]piperazine

  • MDL number MFCD08444723
  • Cas Number : 923208-46-2
  • Molecular Weight : 247.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86639
Availability details for
Quantity
  • Availability
  • Add to Cart