ENA341291445 Enamine

3-(azepane-1-sulfonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08444778
  • Cas Number : 923716-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86675
Availability details for
Quantity
  • Availability
  • Add to Cart