ENA341291448 Enamine

2-[(butylsulfonyl)amino]-3-methylbutanoic acid

  • MDL number MFCD08444782

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86678
Availability details for
Quantity
  • Availability
  • Add to Cart