ENA341291450 Enamine

1-[(4-fluorophenyl)methyl]pyrrolidine-2-carboxylic acid

  • MDL number MFCD08444785
  • Cas Number : 1026725-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86681
Availability details for
Quantity
  • Availability
  • Add to Cart